Learn More
ACROBiosystems Human LILRB2 25ug
Catalog No. 502534509
Encompass_Preferred
Biotinylated Human LILRB2 Protein HisAvitag (LI2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # Q8N423-1).
Catalog No. 50-253-4509 Supplier ACROBiosystems Supplier No. LI2H82E725UG
Biotinylated Human LILRB2 Protein HisAvitag (LI2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # Q8N423-1).