Learn More
Sino Biological Human LILRB2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 14132MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB2 / ILT4 / LIR-2 (rh LILRB2 / ILT4 / LIR-2; Catalog#14132-H08H; AAH36827.1; Met1-Val461). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2316