Learn More
Sino Biological Anti-ILT4/LILRB2 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 14132MM11P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB2 / ILT4 / LIR-2 (rh LILRB2 / ILT4 / LIR-2; Catalog#14132-H08H; AAH36827.1; Met1-Val461) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-9361