Learn More
Sino Biological Human LILRB2 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 14132MM11A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB2/ILT4/LIR-2 (rh LILRB2/ILT4/LIR-2; Catalog#14132-H08H; AAH36827.1; Met1-Val461) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-3362