Learn More
Sino Biological Recombinant Human ILT4/LILRB2 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 14132H49HB20
Encompass_Preferred
A DNA sequence encoding the human LILRB2 (AAH36827.1) (Met1-Val461) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1566