Learn More
Sino Biological Anti-ILT5/LILRB3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11978MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB3 (rh LILRB3; Catalog#11978-H08H; AAI04994.1; Met 1-Glu 443). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1984