Learn More
Sino Biological Anti-ILT5/LILRB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11978RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LILRB3 (rh LILRB3; Catalog#11978-H08H; AAI04994.1; Met 1-Glu 443). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1997