Learn More
Sino Biological Anti-ILT5/LILRB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11978RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LILRB3 (rh LILRB3; Catalog#11978-H08H; AAI04994.1; Met 1-Glu 443). LILRB3 specific IgG was purified by Human LILRB3 affinity chromatography.
Catalog No. 50-240-8387