Learn More
Sino Biological Recombinant Human ILT5/LILRB3 Protein (ECD, His Tag), HPLC-verified, Amine-Labeled Biotinylated
Supplier:  Sino Biological 11978H08HB100
Encompass_Preferred
A DNA sequence encoding the human LILRB3 (NP_006855.2) (Met1-Glu443) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4479