FITC-labeled human LILRB4 (also known as CD85K, ILT3, LIR5, or HM18) is a recombinant protein expressed from HEK293 cells. This protein comprises the extracellular domain of LILRB4 fused to a human IgG1 Fc tag at the C-terminus. It acts as a receptor for class I MHC antigens, recognizing a broad spectrum of HLA-A, HLA-B, HLA-C, and HLA-G alleles, and plays a role in down-regulating immune responses and developing tolerance.
- Excitation wavelength: 488 nm
- Emission wavelength: 535 nm
- Calculated molecular weight: 52.9 kDa
- Purity: >90% as determined by SDS-PAGE
- FITC to protein molar ratio: 1.5–3.5
- Formulation: Lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose protectant
- Storage: Stable at -20°C to -70°C for 12 months (lyophilized) or -70°C for 3 months (reconstituted)