Learn More
Sino Biological Anti-ILT3/LILRB4 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 16742MM05A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB4/CD85k/ILT3 (rh LILRB4/CD85k/ILT3; Catalog#16742-H08H; NP_001265357.2; Met1-Glu259) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0692