Learn More
Sino Biological Recombinant Human ILT3/LILRB4 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 16742H49HB20
Encompass_Preferred
A DNA sequence encoding the Human LILRB4 (NP_001265357.2) (Met1-Glu259) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-0887