Learn More
ACROBiosystems Recombinant Protein;Human LILRB5 / CD85c / LIR-8 Protein;HEK293;100UG;CDC,LILRB5,CD85c,LIR-8,LIR8
Supplier:  ACROBiosystems CDCH5220100UG
Encompass_Preferred
Recombinant Protein;Human LILRB5 / CD85c / LIR-8 Protein;100UG;Human LILRB5, His Tag (CDC-H5220) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 458 (Accession # NP_006831).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 66-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. NC1461381