Learn More
Biomatik Corporation Human LIM And SH3 Protein 1 (LASP1) ELISA Kit
Supplier:  Biomatik Corporation EKU05633
Encompass_Preferred
Human LIM And SH3 Protein 1 (LASP1) ELISA Kit for detecting LASP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14847. Gene ID: 3927. Target Synonyms: MLN50; Metastatic lymph node gene 50 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5595