Learn More
Biomatik Corporation Human LIM Domain Binding Protein 1 (LDB1) ELISA Kit
Supplier:  Biomatik Corporation EKU05634
Encompass_Preferred
Human LIM Domain Binding Protein 1 (LDB1) ELISA Kit for detecting LDB1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86U70. Gene ID: 8861. Target Synonyms: CLIM2; NLI; Carboxy Terminal LIM Domain Protein 2; LIM domain-binding factor CLIM2; Nuclear LIM interactor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4598