Learn More
Biomatik Corporation Human Linker For Activation Of T-Cell (LAT) ELISA Kit
Supplier:  Biomatik Corporation EKU05636
Encompass_Preferred
Human Linker For Activation Of T-Cell (LAT) ELISA Kit for detecting LAT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.102ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43561. Gene ID: 27040. Target Synonyms: LAT1; Pp36; Linker Of Activated T Cells; Linker for activation of T-cells family member 1; 36 kDa phospho-tyrosine adapter protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6403