Learn More
Biomatik Corporation Human Lipase, Bile Salt Dependent (BSDL) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12276
Encompass_Preferred
Human Lipase, Bile Salt Dependent (BSDL) Mini sample ELISA Kit for detecting BSDL in serum, plasma, breast milk, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19835. Target Synonyms: CEL; FAP; BAL; BSSL; FAPP; LIPA; MODY8; Carboxyl Ester Lipase; Bile salt-stimulated lipase; Bucelipase; Cholesterol esterase; Pancreatic lysophospholipase; Sterol esterase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6831