Learn More
Biomatik Corporation Human Lipase Maturation Factor 2 (LMF2) ELISA Kit
Supplier:  Biomatik Corporation EKU08506
Encompass_Preferred
Human Lipase Maturation Factor 2 (LMF2) ELISA Kit for detecting LMF2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BU23. Gene ID: 91289. Target Synonyms: TMEM153; TMEM112B; Transmembrane Protein 153. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6352