Learn More
BIOHIPPO Human LIPB Microsample
SDP
Catalog No. 5000315696
Encompass
Human LIPB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LIPB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LIPB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LIPB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LIPB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15696 Supplier BIOHIPPO Supplier No. BHE15211756
Human LIPB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LIPB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human LIPB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human LIPB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human LIPB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color