Learn More
Biomatik Corporation Human Lipolysis Stimulated Lipoprotein Receptor (LSR) ELISA Kit
Supplier:  Biomatik Corporation EKU05657
Encompass_Preferred
Human Lipolysis Stimulated Lipoprotein Receptor (LSR) ELISA Kit for detecting LSR in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86X29. Gene ID: 51599. Target Synonyms: LISCH7; ILDR3; LISCH; Lipolysis-Stimulated Remnant; Immunoglobulin-Like Domain Containing Receptor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4951