Learn More
American Research Products Inc Human Lipopolysaccharides LPS
Supplier:  American Research Products Inc CSBE09945H
Encompass
Human Lipopolysaccharides LPS ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26813