Learn More
Abcam Human Lipoprotein A Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502585986
Encompass_Preferred
Human Lipoprotein A Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Lipoprotein A.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5986 Supplier Abcam Supplier No. AB24186210X96TESTS
Human Lipoprotein A Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Lipoprotein A.
The product is subject to the following: Abcam Restricted Use Statement