Learn More
Abcam Human Lipoprotein A ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2121651X96TESTS
Encompass_Preferred
Human Lipoprotein A ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human lipoprotein A with a sensitivity of 2.5 ng/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA with a 450 nm readout; compatible with any standard plate reader - Broad sample compatibility – suitable for serum, plasma, urine, and cerebrospinal fluid - Cited in 10 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1843966