Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human lipoProtein(a) native Protein from human plasma

Catalog No. 89950744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-744 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-950-744 Supplier Abnova™ Supplier No. P5332

Used for SDS-PAGE

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation In 10mM NaCl, 0.1mM EDTA, pH 7.2 (0.01% sodium azide)
Name Human lipoProtein(a) native Protein from human plasma
Purification Method Native protein purified from human plasma (from a single polymorph donor) containing EDTA and sodium citrate using ultracentrifugation and affinity chromatography
Quantity 100 μg
Storage Requirements Store at 2-8°C. Do not freeze. Seale under nitrogen or argon after each usage to prevent aggregation and oxidation.
Regulatory Status RUO
Common Name LIPOPROTEIN(A)
Species Human
Form Liquid
Purity or Quality Grade ≥95% by agarose gel electrophoresis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.