Learn More
American Research Products Inc Human lipoprotein associated p
Catalog No. 5000329694
Encompass
Human lipoprotein-associated phospholipase A2 Lp-PLA2 ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29694 Supplier American Research Products Inc Supplier No. CSBE08319H
Human lipoprotein-associated phospholipase A2 Lp-PLA2 ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich