Learn More
ACROBiosystems Human LIV-1 100ug
Supplier:  ACROBiosystems LV1H5257100UG
Encompass_Preferred
Human LIV-1 (229-325) Protein Fc Tag (LV1-H5257) is expressed from human 293 cells (HEK293). It contains AA Lys 229 - Trp 325 (Accession # Q13433-1).
Catalog No. 50-253-4497