Learn More
ACROBiosystems Human LIV-1 200ug
Catalog No. 502534495
Encompass_Preferred
Biotinylated Human LIV-1 Protein FcAvitag (LV1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).
Catalog No. 50-253-4495 Supplier ACROBiosystems Supplier No. LV1H82F5200UG
Biotinylated Human LIV-1 Protein FcAvitag (LV1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).