Learn More
ACROBiosystems Human LIV-1 / SLC39A6 Protein, His Tag
Supplier: ACROBiosystems LV1H5223100UG
Recombinant Protein;100UG;Human LIV-1, His Tag (LV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa.