Learn More
ACROBiosystems Human LIV-1 / SLC39A6 Protein, Fc Tag
Supplier:  ACROBiosystems LV1H52541MG
Encompass_Preferred
Recombinant Protein;1MG;Human LIV-1, Fc Tag (LV1-H5254) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.1 kDa.
Catalog No. 50-205-4932