Learn More
ACROBiosystems ACROBiosystems Human LIV-1 / SLC39A6 Protein, Fc Tag, low endotoxin
Catalog No. 502106277
Encompass_Preferred
Recombinant Protein;1MG;Human LIV-1, Fc Tag (LV1-H5255) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.1 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIV-1, Fc Tag (Cat. No. LV1-H5255) at 1 μg/mL (100 μL/well) can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.6-5 ng/mL (QC tested).
Catalog No. 50-210-6277 Supplier ACROBiosystems Supplier No. LV1H52551MG
Recombinant Protein;1MG;Human LIV-1, Fc Tag (LV1-H5255) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.1 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIV-1, Fc Tag (Cat. No. LV1-H5255) at 1 μg/mL (100 μL/well) can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.6-5 ng/mL (QC tested).