Learn More
ACROBiosystems Human LIV-1 / SLC39A6 Protein, His Tag
Catalog No. 502054930
Encompass_Preferred
Recombinant Protein;1MG;Human LIV-1, His Tag (LV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa.
Catalog No. 50-205-4930 Supplier ACROBiosystems Supplier No. LV1H52231MG
Recombinant Protein;1MG;Human LIV-1, His Tag (LV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa.