Learn More
American Research Products Inc Human LiVer eXpressed antimicr
Catalog No. 5000327444
Encompass
Human LiVer-eXpressed antimicrobial peptide 2 (LEAP2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27444 Supplier American Research Products Inc Supplier No. CSBEL012853HU
Human LiVer-eXpressed antimicrobial peptide 2 (LEAP2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich