Learn More
Biomatik Corporation Human Liver X Receptor Beta (LXRb) ELISA Kit
Supplier:  Biomatik Corporation EKU12208
Encompass_Preferred
Human Liver X Receptor Beta (LXRb) ELISA Kit for detecting LXRb in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55055. Gene ID: 7376. Target Synonyms: NR1-H2; UNR; LXR-B; NER; NER-I; RIP15; Nuclear Receptor Subfamily 1,Group H,Member 2; Ubiquitously-Expressed Nuclear Receptor; Oxysterols receptor LXR-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5499