Learn More
Sino Biological Anti-LMAN2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12455MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LMAN2 (rh LMAN2; Catalog#12455-H08E; Q12907; Asp45-Arg322). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1700