Learn More
Sino Biological Recombinant Human LMAN2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12455H08E1
Encompass_Preferred
A DNA sequence encoding the human LMAN2 extracellular domain (Q12907) (As p45-Arg 322) was fused with a polyhistidine tag at the C-terminus and an initial Met at the N-terminus.
Catalog No. 50-241-0913