Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LMP7/PSMB8 ELISA Kit

Catalog No. EH303RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH303RBX5 5 x 96 Tests
EH303RBX10 10 x 96 Tests
EH303RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH303RBX5 Supplier Invitrogen™ Supplier No. EH303RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human LMP7/PSMB8 quantitates human LMP7/PSMB8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LMP7/PSMB8.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC (major histocompatibility complex). Expression of this gene is induced by gamma interferon and this gene product replaces catalytic subunit 3 (proteasome beta 5 subunit) in the immunoproteasome. Proteolytic processing is required to generate a mature subunit. Two alternative transcripts encoding two isoforms have been identified; both isoforms are processed to yield the same mature subunit.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P28062
Assay Range 2.2-500 ng/mL
Assay Sensitivity 2.2 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; large multifunctional protease 7; Lmp7; Lmp-7; low molecular mass polypeptide 7; low molecular mass protein 7; low molecular weight protein 7; macropain subunit C13; Mc13; MC13 (LMP7); MGC1491; multicatalytic endopeptidase complex subunit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; protease component C13; proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteasome (prosome, macropain) subunit, beta type, 8; proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7); proteasome catalytic subunit 3i; proteasome component C13; proteasome subunit beta 5i; proteasome subunit beta 8; proteasome subunit beta type 8; proteasome subunit beta type-8; Proteasome subunit beta-5i; proteasome subunit MC13; proteasome subunit Y2; proteasome-related gene 7; proteosome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteosome beta 8 subunit; PSMB5i; Psmb8; Really interesting new gene 10 protein; Ring10; Y2ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; large multifunctional protease 7; Lmp7; Lmp-7; low molecular mass polypeptide 7; low molecular mass protein 7; low molecular weight protein 7; macropain subunit C13; Mc13; MC13 (LMP7); MGC1491; multicatalytic endopeptidase complex subunit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; protease component C13; proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteasome (prosome, macropain) subunit, beta type, 8; proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7); proteasome catalytic subunit 3i; proteasome component C13; proteasome subunit beta 5i; proteasome subunit beta 8; proteasome subunit beta type 8; proteasome subunit beta type-8; Proteasome subunit beta-5i; proteasome subunit MC13; proteasome subunit Y2; proteasome-related gene 7; proteosome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteosome beta 8 subunit; PSMB5i; Psmb8; Really interesting new gene 10 protein; Ring10; Y2;
Gene ID (Entrez) 5696
Gene Symbol Psmb8
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PSMB8
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.