Learn More
American Research Products Inc Human Low affinity immunoglobu
Catalog No. 5000328260
Encompass
Human Low affinity immunoglobulin gamma Fc region receptor III-B (FCGR3B) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28260 Supplier American Research Products Inc Supplier No. CSBEL008544HU
Human Low affinity immunoglobulin gamma Fc region receptor III-B (FCGR3B) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich