Learn More
Biomatik Corporation Human Low Density Lipoprotein Receptor Related Protein 1B (LRP1B) ELISA Kit
Supplier:  Biomatik Corporation EKU11051
Encompass_Preferred
Human Low Density Lipoprotein Receptor Related Protein 1B (LRP1B) ELISA Kit for detecting LRP1B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.47-30ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NZR2. Gene ID: 53353. Target Synonyms: LRPDIT; LRP-DIT (deleted In Tumors) ; Low-density lipoprotein receptor-related protein-deleted in tumor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4701