Learn More
Sino Biological Anti-LOXL2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11664MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LOXL2 / Lysyl oxidase-like 2 (rh LOXL2 / Lysyl oxidase-like 2; Catalog#11664-H08H; Q9Y4K0; Met1-Gln774). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4641