Learn More
ACROBiosystems ACROBiosystems Human LOXL2 Protein, His Tag
Supplier:  ACROBiosystems LO2H52H3500UG
Encompass_Preferred
Recombinant Protein;500UG;Human LOXL2, His Tag (LO2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession # Q9Y4K0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 87.7 kDa.
Catalog No. 50-210-6226