Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Lp-PLA2/PLA2G7/PAF-AH ELISA Kit

Catalog No. EH304RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH304RBX5 5 x 96 Tests
EH304RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH304RBX5 Supplier Invitrogen™ Supplier No. EH304RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Lp-PLA2/PLA2G7/PAF-AH quantitates human Lp-PLA2/PLA2G7/PAF-AH in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Lp-PLA2/PLA2G7/PAF-AH.

Platelet Activating Factor (PAF) is a biologically active phospholipid, which exerts primarily proinflammatory activities by specifically signaling through G-protein-coupled receptors on platelets, neutrophils, and monocytes. Platelet Activating Factor Acetylhydrolase (PAF-AH) is a secreted protein that mediates PAF activity by specifically catalyzing hydrolysis of the 'sn2' ester bond, resulting in the conversion of PAF to the biologically inactive lyso-PAF. PAF-AH can also interact with LDL particles to induce the hydrolysis of LDL-associated, oxidized phospholipids, generating lysophosphatidylcholine (lyso-PC) and other lysophospholipids.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13093
Assay Range 8.19-2000 ng/mL
Assay Sensitivity 8.1 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1-alkyl-2-acetylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase; Group-VIIA phospholipase A2; gVIIA-PLA2; LDL-associated phospholipase A2; LDL-PLA(2); LDL-PLA2; lipoprotein-associated phospholipase A2; LP-PLA2; PAF 2-acylhydrolase; PAF acetylhydrolase; PAFAD; Pafah; phospholipase A2 group VII; phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma); PLA2G7; Platelet-activating factor acetylhydrolase; R754001-alkyl-2-acetylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase; Group-VIIA phospholipase A2; gVIIA-PLA2; LDL-associated phospholipase A2; LDL-PLA(2); LDL-PLA2; lipoprotein-associated phospholipase A2; LP-PLA2; PAF 2-acylhydrolase; PAF acetylhydrolase; PAFAD; Pafah; phospholipase A2 group VII; phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma); PLA2G7; Platelet-activating factor acetylhydrolase; R75400;
Gene ID (Entrez) 7941
Gene Symbol PLA2G7
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PLA2G7
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.