Learn More
Biomatik Corporation Human LPA (Lipoprotein a) ELISA Kit
Supplier:  Biomatik Corporation EKF58628
Encompass_Preferred
Human LPA (Lipoprotein a) ELISA Kit for detecting LPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.234-15ng/ml. Sensitivity: 0.141ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08519. Target Synonyms: LP-a; LPA; Lp; a; AK38; APOA; LP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7429