Learn More
ACROBiosystems Human LPAR1 Full Length Protein (VLP) 100ug
Catalog No. NC3880896
Encompass_Preferred
Source:Human LPAR1 Full Length Protein (VLP) (LP1-H52P3) is expressed from human 293 cells (HEK293). It contains AA Ala 2 - Val 364 (Accession # Q92633).Predicted N-terminus: Asp
Catalog No. NC3880896 Supplier ACROBiosystems Supplier No. LP1H52P3100UG
Source:Human LPAR1 Full Length Protein (VLP) (LP1-H52P3) is expressed from human 293 cells (HEK293). It contains AA Ala 2 - Val 364 (Accession # Q92633).Predicted N-terminus: Asp