Learn More
Immunology Consultants Human LRG1 ELISA Kit
SDP
Supplier:  Immunology Consultants E80LRG
Encompass_Preferred
The Human LRG1 test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for the quantitative determination of Human leucine-rich alpha-2 glycoprotein 1 (LRG1) in biological samples.
Catalog No. 50-240-6626