Learn More
Sino Biological Human LRG1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 13371R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LRG1 (rh LRG1; Catalog#13371-H08H; AAH34389.1; Met1-Gln347).
Catalog No. 50-237-3713