Learn More
Sino Biological Anti-LRG1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13371T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LRG1 (rh LRG1; Catalog#13371-H08H; AAH34389.1; Met1-Gln347). LRG1 specific IgG was purified by Human LRG1 affinity chromatography.
Catalog No. 50-255-4247