Learn More
Raybiotech Inc Human LRIG3 ELISA Kit
Supplier:  Raybiotech Inc ELHLRIG31
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2.4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human LRIG3.
Catalog No. 50-194-9509