Learn More
Invitrogen™ Human LRIG3 ELISA Kit
Description
Human LRIG3 quantitates human LRIG3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LRIG3.
Specifications
Specifications
| Accession Number | Q6UXM1 |
| Assay Range | 2.4-500 ng/mL |
| Assay Sensitivity | 2.4 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | leucine rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin-like domains 3; leucine-rich repeats and immunoglobulin-like domains protein 3; LIG3; LIG-3; LRIG3; RGD1561255; UNQ287/PRO326/PRO335leucine rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin-like domains 3; leucine-rich repeats and immunoglobulin-like domains protein 3; LIG3; LIG-3; LRIG3; RGD1561255; UNQ287/PRO326/PRO335; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.