Learn More
ACROBiosystems Human LRP-1 / cluster Ⅲ Protein, Fc Tag
Catalog No. 502331573
Encompass_Preferred
Human cluster Ⅲ Protein, Fc Tag (LR1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 2522 - Ile 2941 (Accession # Q07954-1).
Catalog No. 50-233-1573 Supplier ACROBiosystems Supplier No. LR1H5255100UG
Human cluster Ⅲ Protein, Fc Tag (LR1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 2522 - Ile 2941 (Accession # Q07954-1).