Learn More
ACROBiosystems Human LRP-1 / cluster Ⅳ Protein, Fc Tag
Catalog No. 502331581
Encompass_Preferred
Human cluster Ⅳ Protein, Fc Tag (LR1-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 3332 - Asp 3779 (Accession # Q07954-1).
Catalog No. 50-233-1581 Supplier ACROBiosystems Supplier No. LR1H52561MG
Human cluster Ⅳ Protein, Fc Tag (LR1-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 3332 - Asp 3779 (Accession # Q07954-1).